siRNA Details
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 19 | 2360 | ||
NM_001162501.1 | NP_001155973.1 | 18 | 4681 | |||
NM_015088.2 | NP_055903.2 | 16 | 4351 | |||
GenBank mRNA | AB029016.2 | - | 16 | 4331 | ||
AK294519.1 | - | 18 | 4672 | |||
AK296590.1 | - | 9 | 1400 | |||
AK304087.1 | - | 13 | 1836 | |||
BC028626.2 | - | 19 | 2293 | 4826787 | ||
BX647630.1 | - | 13 | 3553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
