siRNA Details
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 10 | 1822 | ||
NM_015264.1 | NP_056079.1 | 10 | 1894 | |||
XM_011530017.2 | XP_011528319.1 | 10 | 1857 | |||
XM_017028672.1 | XP_016884161.1 | 10 | 1866 | |||
GenBank mRNA | AB023147.1 | - | 9 | 1590 | ||
AK025608.1 | - | 8 | 1411 | |||
AK094359.1 | - | 1 | 1348 | |||
AK094937.1 | - | 11 | 1905 | |||
AK124648.1 | - | 10 | 1902 | |||
AY007105.1 | - | 3 | 776 | |||
BC016882.1 | - | 5 | 1152 | |||
BC038414.1 | - | 9 | 1969 | |||
BC053595.1 | - | 10 | 1816 | 5730020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
