siRNA Details
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 9, 10 | 1235 | ||
NM_015264.1 | NP_056079.1 | 9, 10 | 1307 | |||
XM_011530017.2 | XP_011528319.1 | 9, 10 | 1270 | |||
XM_017028672.1 | XP_016884161.1 | 9, 10 | 1279 | |||
GenBank mRNA | AB023147.1 | - | 8, 9 | 1003 | ||
AK025608.1 | - | 7, 8 | 824 | |||
AK094937.1 | - | 10, 11 | 1318 | |||
AK124648.1 | - | 9, 10 | 1315 | |||
AK297058.1 | - | 9, 10 | 1287 | |||
AK309015.1 | - | 8, 9 | 1359 | |||
AK315883.1 | - | 9, 10 | 1219 | |||
AY007105.1 | - | 2, 3 | 189 | |||
BC016882.1 | - | 4, 5 | 565 | |||
BC038414.1 | - | 8, 9 | 1382 | |||
BC053595.1 | - | 9, 10 | 1229 | 5730020 | ||
CR456402.1 | - | 9, 10 | 1172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
