siRNA Details
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 9 | 1206 | ||
NM_015264.1 | NP_056079.1 | 9 | 1278 | |||
XM_011530017.2 | XP_011528319.1 | 9 | 1241 | |||
XM_017028672.1 | XP_016884161.1 | 9 | 1250 | |||
GenBank mRNA | AB023147.1 | - | 8 | 974 | ||
AK025608.1 | - | 7 | 795 | |||
AK094359.1 | - | 1 | 159 | |||
AK094937.1 | - | 10 | 1289 | |||
AK124648.1 | - | 9 | 1286 | |||
AK297058.1 | - | 9 | 1258 | |||
AK309015.1 | - | 8 | 1330 | |||
AK315883.1 | - | 9 | 1190 | |||
AY007105.1 | - | 2 | 160 | |||
BC016882.1 | - | 4 | 536 | |||
BC038414.1 | - | 8 | 1353 | |||
BC053595.1 | - | 9 | 1200 | 5730020 | ||
CR456402.1 | - | 9 | 1143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
