siRNA Details
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267571.1 | NP_001254500.1 | 13 | 3275 | ||
NM_001267572.1 | NP_001254501.1 | 7 | 2054 | |||
NM_018421.3 | NP_060891.3 | 13 | 3242 | |||
NR_051978.1 | 12 | 3133 | ||||
XM_011518843.1 | XP_011517145.1 | 8 | 3157 | |||
XM_017014899.1 | XP_016870388.1 | 13 | 2705 | |||
GenBank mRNA | AF318370.1 | - | 13 | 3130 | ||
AK026105.1 | - | 12 | 2623 | |||
AK124772.1 | - | 12 | 3824 | |||
AK131290.1 | - | 10 | 2531 | |||
AL353935.1 | - | 8 | 3336 | |||
AY026527.1 | - | 13 | 3154 | |||
BC028918.1 | - | 7 | 2054 | 4931447 | ||
BC071978.1 | - | 9 | 2162 | 5443205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
