siRNA Details
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168368.1 | NP_001161840.1 | 15 | 4258 | ||
NM_020692.2 | NP_065743.2 | 16 | 3097 | |||
XM_017021498.1 | XP_016876987.1 | 17 | 3084 | |||
GenBank mRNA | AB032956.1 | - | 15 | 5504 | ||
AK091426.1 | - | 1 | 2391 | |||
BC036812.1 | - | 16 | 2848 | 5734009 | ||
BC098578.1 | - | 16 | 2863 | 30924466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
