siRNA Details
Gene Symbol: KIAA0368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080398.1 | NP_001073867.1 | 13, 14 | 1751 | ||
XM_005251850.3 | XP_005251907.1 | 11, 12 | 1254 | |||
XM_005251851.4 | XP_005251908.1 | 12, 13 | 1633 | |||
XM_005251852.4 | XP_005251909.1 | 12, 13 | 1585 | |||
XM_005251853.3 | XP_005251910.1 | 12, 13 | 1367 | |||
XM_005251854.3 | XP_005251911.1 | 11, 12 | 1536 | |||
XM_011518422.2 | XP_011516724.1 | 11, 12 | 1254 | |||
GenBank mRNA | AB002366.1 | - | 1, 2 | 25 | ||
AK122712.1 | - | 11, 12 | 1238 | |||
AK127247.1 | - | 7, 8 | 694 | |||
AK308313.1 | - | 12, 13 | 1358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
