siRNA Details
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 25 | 2697 | ||
XM_011528481.2 | XP_011526783.1 | 25 | 2725 | |||
GenBank mRNA | AB014511.1 | - | 20 | 2251 | ||
AK172802.1 | - | 23 | 2386 | |||
AK299064.1 | - | 25 | 2626 | |||
BC110591.1 | - | 21 | 2189 | |||
BC110592.1 | - | 21 | 2189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
