siRNA Details
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 28 | 3246 | ||
XM_011528481.2 | XP_011526783.1 | 28 | 3274 | |||
GenBank mRNA | AB014511.1 | - | 23 | 2800 | ||
AK172802.1 | - | 26 | 2936 | |||
AK299064.1 | - | 28 | 3175 | |||
BC110591.1 | - | 24 | 2738 | |||
BC110592.1 | - | 24 | 2738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
