siRNA Details
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 3 | 345 | ||
XM_011528481.2 | XP_011526783.1 | 3 | 373 | |||
GenBank mRNA | AB014511.1 | - | 3 | 262 | ||
AK172802.1 | - | 1 | 34 | |||
AK299064.1 | - | 3 | 274 | |||
HY147052.1 | - | 3 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
