siRNA Details
Gene Symbol: BTF3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136497.2 | NP_001129969.1 | 5 | 768 | ||
NM_001243767.1 | NP_001230696.1 | 5 | 633 | |||
NM_152265.4 | NP_689478.1 | 6 | 835 | |||
GenBank mRNA | AK027582.1 | - | 5 | 649 | ||
AK027750.1 | - | 6 | 717 | |||
BC021004.1 | - | 5 | 612 | |||
BC022371.1 | - | 6 | 697 | 4739327 | ||
BC070378.1 | - | 6 | 725 | 6495112 | ||
BU785869.1 | - | 5 | 477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
