siRNA Details
Gene Symbol: C5orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023073.3 | NP_075561.3 | 5 | 621 | ||
XM_005248345.3 | XP_005248402.1 | 5 | 609 | |||
XM_005248346.3 | XP_005248403.1 | 5 | 529 | |||
XM_005248347.3 | XP_005248404.1 | 5 | 529 | |||
XM_005248349.3 | XP_005248406.1 | 5 | 529 | |||
XM_005248350.3 | XP_005248407.1 | 5 | 527 | |||
XM_011514085.2 | XP_011512387.1 | 5 | 528 | |||
XM_011514086.2 | XP_011512388.1 | 5 | 525 | |||
XM_011514087.2 | XP_011512389.1 | 5 | 614 | |||
XM_011514088.2 | XP_011512390.1 | 5 | 614 | |||
XM_011514089.2 | XP_011512391.1 | 5 | 614 | |||
XM_011514090.2 | XP_011512392.1 | 5 | 392 | |||
XM_011514092.2 | XP_011512394.1 | 5 | 614 | |||
XM_017009760.1 | XP_016865249.1 | 4 | 340 | |||
XM_017009761.1 | XP_016865250.1 | 4 | 308 | |||
XM_017009762.1 | XP_016865251.1 | 2 | 174 | |||
GenBank mRNA | DW009948.1 | - | 3 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
