siRNA Details
Gene Symbol: DDX12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033399.1 | 8 | 1016 | |||
GenBank mRNA | U33834.1 | - | 5 | 377 |
Gene Symbol: LOC642846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024374.1 | 8 | 1065 | |||
GenBank mRNA | AK296420.1 | - | 8 | 1073 | ||
AK303618.1 | - | 8 | 843 | |||
AK309820.1 | - | 9 | 1005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
