siRNA Details
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256539.3 | NP_001243468.2 | 2 | 536 | ||
NM_001256540.3 | NP_001243469.2 | 2 | 536 | |||
NM_001291305.2 | NP_001278234.1 | 2 | 536 | |||
NM_001291307.2 | NP_001278236.1 | 2 | 536 | |||
NM_001291308.2 | NP_001278237.1 | 2 | 536 | |||
NM_001317975.1 | NP_001304904.1 | 1 | 625 | |||
NM_016391.7 | NP_057475.2 | 2 | 536 | |||
NR_134316.1 | 1 | 625 | ||||
NR_134318.1 | 2 | 536 | ||||
XM_011534567.1 | XP_011532869.1 | 2 | 215 | |||
GenBank mRNA | AF151019.1 | - | 2 | 534 | ||
AF151875.1 | - | 3 | 604 | |||
AF161460.1 | - | 2 | 210 | |||
AK300883.1 | - | 2 | 215 | |||
AK303281.1 | - | 2 | 207 | |||
AW837047.1 | - | 1 | 274 | |||
AY561704.1 | - | 2 | 148 | |||
BC019331.1 | - | 2 | 371 | 3534019 | ||
BC032424.2 | - | 2 | 179 | 5416724 | ||
BC040106.1 | - | 2 | 196 | 6066208 | ||
BM826387.1 | - | 2 | 195 | |||
BP234677.1 | - | 1 | 296 | |||
BU594763.1 | - | 2 | 210 | |||
CB122385.1 | - | 2 | 196 | |||
CR457169.1 | - | 2 | 148 | |||
HY064071.1 | - | 2 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
