siRNA Details
Gene Symbol: DCAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284206.1 | NP_001271135.1 | 6 | 937 | ||
NM_001284207.1 | NP_001271136.1 | 6 | 693 | |||
NM_001284208.1 | NP_001271137.1 | 6 | 940 | |||
NM_003861.2 | NP_003852.1 | 6 | 940 | |||
XM_006720297.2 | XP_006720360.1 | 6 | 940 | |||
XM_006720298.2 | XP_006720361.1 | 6 | 937 | |||
XM_006720299.3 | XP_006720362.1 | 6 | 575 | |||
XM_006720300.2 | XP_006720363.1 | 6 | 596 | |||
XM_011537278.1 | XP_011535580.1 | 7 | 1209 | |||
XM_011537279.2 | XP_011535581.1 | 6 | 672 | |||
XM_011537280.2 | XP_011535582.1 | 2 | 163 | |||
XM_017021733.1 | XP_016877222.1 | 6 | 530 | |||
XM_017021734.1 | XP_016877223.1 | 6 | 592 | |||
XM_017021735.1 | XP_016877224.1 | 6 | 672 | |||
XM_017021736.1 | XP_016877225.1 | 6 | 596 | |||
XM_017021737.1 | XP_016877226.1 | 2 | 225 | |||
XM_017021738.1 | XP_016877227.1 | 6 | 548 | |||
XM_017021739.1 | XP_016877228.1 | 6 | 635 | |||
GenBank mRNA | AB058727.1 | - | 6 | 775 | ||
AL831932.2 | - | 6 | 693 | |||
BC022967.1 | - | 6 | 727 | |||
BC136632.1 | - | 6 | 869 | 9020622 | ||
BX648006.1 | - | 6 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
