siRNA Details
Gene Symbol: DCAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284208.1 | NP_001271137.1 | 7 | 1985 | ||
XM_017021738.1 | XP_016877227.1 | 7 | 1593 | |||
XM_017021739.1 | XP_016877228.1 | 7 | 1680 | |||
GenBank mRNA | BC022967.1 | - | 7 | 1772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
