siRNA Details
Gene Symbol: EARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083614.1 | NP_001077083.1 | 9 | 3222 | ||
NR_003501.1 | 10 | 3129 | ||||
XM_011545738.1 | XP_011544040.1 | 9 | 3512 | |||
XM_011545739.1 | XP_011544041.1 | 9 | 3131 | |||
GenBank mRNA | AB075850.1 | - | 10 | 3086 | ||
AK095998.1 | - | 10 | 2733 | |||
AL832489.1 | - | 10 | 3099 | |||
BX648129.1 | - | 8 | 2802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
