siRNA Details
Gene Symbol: NAA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011713.2 | NP_001011713.2 | 3 | 951 | ||
GenBank mRNA | AK092674.1 | - | 3 | 951 | ||
AK296372.1 | - | 2 | 188 | |||
AK304257.1 | - | 3 | 555 | |||
AK307348.1 | - | 2 | 144 | |||
BC118589.1 | - | 3 | 906 | 40085064 | ||
BC122557.1 | - | 4 | 795 | 40085054 | ||
BX648700.1 | - | 2 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
