siRNA Details
Gene Symbol: ZNF835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005850.2 | NP_001005850.2 | 2 | 394 | ||
XM_005259382.2 | XP_005259439.1 | 2 | 397 | |||
XM_005259383.3 | XP_005259440.1 | 2 | 429 | |||
GenBank mRNA | AK299565.1 | - | 2 | 394 | ||
DC382896.1 | - | 2 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
