siRNA Details
Gene Symbol: EBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110514.1 | NP_001103984.1 | 17, 18 | 2067 | ||
XM_006723600.1 | XP_006723663.1 | 17, 18 | 1862 | |||
XM_017027983.1 | XP_016883472.1 | 18, 19 | 1959 | |||
XM_017027984.1 | XP_016883473.1 | 18, 19 | 2065 | |||
XM_017027985.1 | XP_016883474.1 | 11, 12 | 1221 | |||
GenBank mRNA | AB037863.1 | - | 17, 18 | 1945 | ||
BC019106.2 | - | 3, 4 | 558 | |||
BC037555.1 | - | 9, 10 | 1167 | |||
BC054347.1 | - | 2, 3 | 721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
