siRNA Details
Gene Symbol: RASAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022904.1 | NP_075055.1 | 11 | 1795 | ||
GenBank mRNA | AK024488.1 | - | 10 | 2240 | ||
AK122587.1 | - | 11 | 1790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
