siRNA Details
Gene Symbol: RASAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022904.1 | NP_075055.1 | 18 | 2992 | ||
XM_011528185.1 | XP_011526487.1 | 18 | 3001 | |||
XM_011528186.1 | XP_011526488.1 | 18 | 2983 | |||
XM_011528187.1 | XP_011526489.1 | 17 | 2937 | |||
GenBank mRNA | AK024488.1 | - | 16 | 3989 | ||
AK122587.1 | - | 18 | 2987 | |||
AL832868.1 | - | 4 | 449 | |||
BC030281.1 | - | 5 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
