siRNA Details
Gene Symbol: TBX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152380.2 | NP_689593.2 | 8 | 1313 | ||
XM_005271161.3 | XP_005271218.1 | 9 | 1868 | |||
XM_005271162.1 | XP_005271219.1 | 8 | 1769 | |||
XM_011542052.2 | XP_011540354.1 | 5 | 814 | |||
GenBank mRNA | AK096396.1 | - | 4 | 600 | ||
AK127536.1 | - | 8 | 1313 | |||
BC122553.1 | - | 8 | 1131 | 40077247 | ||
BX537778.1 | - | 3 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
