siRNA Details
Gene Symbol: FAM171A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010924.1 | NP_001010924.1 | 2 | 323 | ||
XM_011519378.2 | XP_011517680.1 | 2 | 983 | |||
XM_017015904.1 | XP_016871393.1 | 2 | 244 | |||
XM_017015905.1 | XP_016871394.1 | 1 | 17 | |||
GenBank mRNA | AL050367.1 | - | 2 | 323 | ||
AY683003.1 | - | 2 | 370 | |||
BC109302.1 | - | 2 | 323 | 40035211 | ||
BC109303.1 | - | 2 | 323 | 40035213 | ||
BX496483.1 | - | 2 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
