siRNA Details
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308068.1 | NP_001294997.1 | 10 | 4278 | ||
NM_032296.2 | NP_115672.2 | 10 | 4275 | |||
GenBank mRNA | AB046772.1 | - | 10 | 4255 | ||
BC028572.1 | - | 10 | 4279 | 4839062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
