siRNA Details
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308068.1 | NP_001294997.1 | 10 | 2905 | ||
NM_032296.2 | NP_115672.2 | 10 | 2902 | |||
XM_006720962.3 | XP_006721025.1 | 12 | 3154 | |||
XM_011522701.2 | XP_011521003.1 | 11 | 3543 | |||
XM_011522702.2 | XP_011521004.1 | 11 | 4017 | |||
XM_011522703.2 | XP_011521005.1 | 12 | 3638 | |||
XM_011522704.2 | XP_011521006.1 | 11 | 3541 | |||
XM_011522705.2 | XP_011521007.1 | 12 | 3299 | |||
XM_017023772.1 | XP_016879261.1 | 13 | 3735 | |||
GenBank mRNA | AB046772.1 | - | 10 | 2882 | ||
AK026885.1 | - | 5 | 1071 | |||
AK055777.1 | - | 4 | 1667 | |||
AK057022.1 | - | 5 | 2382 | |||
AK092434.1 | - | 6 | 1722 | |||
AK127235.1 | - | 5 | 3026 | |||
AL136585.1 | - | 8 | 1874 | |||
BC018712.1 | - | 6 | 1650 | 4875281 | ||
BC028572.1 | - | 10 | 2906 | 4839062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
