siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 27, 28 | 5074 | ||
XM_011544278.2 | XP_011542580.2 | 20, 21 | 2685 | |||
XM_017002391.1 | XP_016857880.1 | 26, 27 | 4638 | |||
XM_017002392.1 | XP_016857881.1 | 14, 15 | 1924 | |||
GenBank mRNA | AB007895.1 | - | 4, 5 | 1990 | ||
AK055374.1 | - | 8, 9 | 1240 | |||
AK074078.1 | - | 22, 23 | 4146 | |||
AK090784.1 | - | 3, 4 | 1306 | |||
AK097229.1 | - | 13, 14 | 1649 | |||
AK307812.1 | - | 14, 15 | 1919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
