siRNA Details
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 20 | 2329 | ||
NM_001291721.1 | NP_001278650.1 | 19 | 2251 | |||
NM_001291722.1 | NP_001278651.1 | 21 | 2404 | |||
NM_014376.3 | NP_055191.2 | 20 | 2455 | |||
XM_011534516.2 | XP_011532818.1 | 20 | 2595 | |||
XM_017009341.1 | XP_016864830.1 | 20 | 2444 | |||
XM_017009342.1 | XP_016864831.1 | 21 | 2366 | |||
GenBank mRNA | AB032994.2 | - | 21 | 2344 | ||
AF160973.1 | - | 20 | 2238 | |||
AK223604.1 | - | 20 | 2282 | |||
AK294220.1 | - | 15 | 1698 | |||
AK295865.1 | - | 17 | 1878 | |||
AK297401.1 | - | 18 | 2061 | |||
AK303702.1 | - | 19 | 2204 | |||
AL136549.1 | - | 20 | 2328 | |||
AL161999.1 | - | 12 | 1289 | |||
BC011762.1 | - | 20 | 2455 | 3619680 | ||
BC021008.1 | - | 20 | 2455 | |||
L47738.1 | - | 9 | 1049 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
