siRNA Details
Gene Symbol: MUC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002458.2 | NP_002449.2 | 31 | 7401 | ||
GenBank mRNA | AF253321.1 | - | 2 | 1570 | ||
U63836.1 | - | 1 | 442 | |||
U78550.1 | - | 1 | 34 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
