siRNA Details
Gene Symbol: MUC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002458.2 | NP_002449.2 | 11 | 1378 | ||
GenBank mRNA | AF086604.1 | - | 11 | 1316 | ||
AK310939.1 | - | 11 | 1377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
