siRNA Details
Gene Symbol: ZFP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320666.1 | NP_001307595.1 | 6 | 1785 | ||
NM_001320667.1 | NP_001307596.1 | 6 | 1691 | |||
NM_001320668.1 | NP_001307597.1 | 6 | 1561 | |||
NM_001320669.1 | NP_001307598.1 | 6 | 1499 | |||
NM_001320670.1 | NP_001307599.1 | 6 | 1496 | |||
NM_001320671.1 | NP_001307600.1 | 6 | 1402 | |||
NM_014898.2 | NP_055713.1 | 6 | 1732 | |||
GenBank mRNA | AB023178.1 | - | 6 | 1732 | ||
AK297301.1 | - | 7 | 1118 | |||
BC041087.1 | - | 6 | 1761 | 5579247 | ||
BC050542.1 | - | 6 | 1690 | |||
GU388430.1 | - | 6 | 1561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
