siRNA Details
Gene Symbol: IAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039613.2 | NP_001034702.1 | 5 | 513 | ||
NM_001320858.1 | NP_001307787.1 | 5 | 613 | |||
NM_001320859.1 | NP_001307788.1 | 4 | 460 | |||
NM_001320860.1 | NP_001307789.1 | 4 | 428 | |||
NM_001320863.1 | NP_001307792.1 | 5 | 481 | |||
XM_017003878.1 | XP_016859367.1 | 4 | 603 | |||
GenBank mRNA | AK092982.1 | - | 4 | 425 | ||
AK291984.1 | - | 5 | 480 | |||
AK297648.1 | - | 4 | 414 | |||
BC019061.1 | - | 4 | 452 | |||
BC039709.1 | - | 5 | 480 | |||
BC059410.1 | - | 4 | 435 | |||
BC067865.1 | - | 4 | 424 | |||
BC111025.1 | - | 5 | 501 | 5200514 | ||
BF447213.1 | - | 5 | 613 | |||
BM553525.1 | - | 5 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
