siRNA Details
Gene Symbol: STAG3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040585.1 | 6 | 1166 | |||
NR_040586.1 | 5 | 1090 | ||||
GenBank mRNA | AK023257.1 | - | 5 | 1084 | ||
AK308931.1 | - | 6 | 948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
