siRNA Details
Gene Symbol: TMEM205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145416.1 | NP_001138888.1 | 3 | 330 | ||
NM_001321112.1 | NP_001308041.1 | 3 | 439 | |||
NM_001321113.1 | NP_001308042.1 | 3 | 249 | |||
NM_001321114.1 | NP_001308043.1 | 3 | 358 | |||
NM_033408.3 | NP_212133.1 | 3 | 449 | |||
NM_198536.2 | NP_940938.1 | 2 | 828 | |||
XM_005259899.2 | XP_005259956.1 | 3 | 535 | |||
XM_017026769.1 | XP_016882258.1 | 1 | 140 | |||
GenBank mRNA | AK127147.1 | - | 2 | 2715 | ||
AK291979.1 | - | 3 | 309 | |||
AY358949.1 | - | 3 | 270 | |||
BC064948.1 | - | 2 | 569 | 6025162 | ||
BC091472.1 | - | 3 | 257 | 30335220 | ||
BE889078.1 | - | 3 | 449 | |||
BM352837.1 | - | 3 | 208 | |||
BM451378.1 | - | 3 | 335 | |||
BQ639667.1 | - | 3 | 366 | |||
CB994930.1 | - | 3 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
