siRNA Details
Gene Symbol: OLFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288821.1 | NP_001275750.1 | 4 | 703 | ||
NM_001288823.1 | NP_001275752.1 | 5 | 1015 | |||
NM_058170.3 | NP_477518.2 | 4 | 954 | |||
NR_110210.1 | 5 | 1015 | ||||
NR_110211.1 | 4 | 954 | ||||
NR_110212.1 | 4 | 703 | ||||
XM_017000240.1 | XP_016855729.1 | 5 | 858 | |||
GenBank mRNA | AF397392.2 | - | 4 | 596 | ||
AF397393.2 | - | 4 | 596 | |||
AF397394.1 | - | 4 | 625 | |||
AF397395.1 | - | 4 | 625 | |||
AF397396.1 | - | 5 | 686 | |||
AF397397.1 | - | 5 | 686 | |||
AK095551.1 | - | 2 | 180 | |||
AK095724.1 | - | 4 | 750 | |||
AK300870.1 | - | 4 | 703 | |||
AY358722.1 | - | 4 | 667 | |||
BC022531.1 | - | 4 | 712 | 4812035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
