siRNA Details
Gene Symbol: SF3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006842.2 | NP_006833.2 | 16 | 1978 | ||
XM_005273726.3 | XP_005273783.1 | 16 | 2548 | |||
XM_011544740.2 | XP_011543042.1 | 16 | 2548 | |||
XM_017017144.1 | XP_016872633.1 | 16 | 2545 | |||
GenBank mRNA | AK290850.1 | - | 16 | 1962 | ||
AK300016.1 | - | 10 | 1252 | |||
BC000401.2 | - | 16 | 1937 | |||
BC007610.1 | - | 9 | 1161 | 3351485 | ||
BC014125.2 | - | 4 | 366 | |||
BC053577.1 | - | 16 | 1945 | |||
BX537771.1 | - | 14 | 1651 | |||
U41371.1 | - | 16 | 1917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
