siRNA Details
Gene Symbol: SF3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006842.2 | NP_006833.2 | 13 | 1505 | ||
XM_005273726.3 | XP_005273783.1 | 13 | 2075 | |||
XM_011544740.2 | XP_011543042.1 | 13 | 2075 | |||
XM_017017144.1 | XP_016872633.1 | 13 | 2072 | |||
GenBank mRNA | AK290850.1 | - | 13 | 1489 | ||
AK300016.1 | - | 7 | 779 | |||
BC000401.2 | - | 13 | 1464 | |||
BC007610.1 | - | 6 | 688 | 3351485 | ||
BC053577.1 | - | 13 | 1472 | |||
BX537771.1 | - | 11 | 1178 | |||
U41371.1 | - | 13 | 1444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
