siRNA Details
Gene Symbol: CENPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127181.2 | NP_001120653.1 | 6 | 1319 | ||
NM_001171182.1 | NP_001164653.1 | 4 | 792 | |||
NM_033319.3 | NP_201576.1 | 5 | 1181 | |||
GenBank mRNA | AK055606.1 | - | 909 | |||
BC007071.1 | - | 5 | 716 | 3682908 | ||
BC019022.2 | - | 4 | 793 | |||
BC033154.1 | - | 6 | 1002 | 4139038 | ||
BC066658.1 | - | 5 | 863 | 6172418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
