siRNA Details
Gene Symbol: FAM149A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006655.2 | NP_001006656.1 | 13 | 1706 | ||
NM_015398.2 | NP_056213.1 | 14 | 2005 | |||
XM_005262911.1 | XP_005262968.1 | 14 | 1937 | |||
XM_005262912.1 | XP_005262969.1 | 14 | 2008 | |||
XM_005262913.1 | XP_005262970.1 | 14 | 1845 | |||
XM_011531834.1 | XP_011530136.1 | 13 | 1701 | |||
XM_011531835.2 | XP_011530137.1 | 14 | 1963 | |||
XM_017007996.1 | XP_016863485.1 | 14 | 2522 | |||
XM_017007997.1 | XP_016863486.1 | 14 | 2548 | |||
XM_017007999.1 | XP_016863488.1 | 14 | 3309 | |||
XM_017008000.1 | XP_016863489.1 | 14 | 1793 | |||
XM_017008001.1 | XP_016863490.1 | 14 | 1882 | |||
XM_017008002.1 | XP_016863491.1 | 14 | 1966 | |||
XM_017008003.1 | XP_016863492.1 | 14 | 1822 | |||
XM_017008004.1 | XP_016863493.1 | 14 | 1874 | |||
XM_017008005.1 | XP_016863494.1 | 14 | 1979 | |||
XM_017008006.1 | XP_016863495.1 | 14 | 1871 | |||
GenBank mRNA | AF173894.1 | - | 13 | 1642 | ||
AK057166.1 | - | 14 | 1835 | |||
AL080065.1 | - | 8 | 1051 | |||
BC110816.1 | - | 14 | 1907 | |||
BC136792.1 | - | 11 | 1451 | 9020782 | ||
BC142992.1 | - | 14 | 1959 | 8860446 | ||
BC144411.1 | - | 11 | 1448 | 9052932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
