siRNA Details
Gene Symbol: FAM168A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286050.1 | NP_001272979.1 | 7 | 817 | ||
NM_015159.2 | NP_055974.1 | 6 | 790 | |||
XM_017017415.1 | XP_016872904.1 | 4 | 397 | |||
GenBank mRNA | AK292054.1 | - | 6 | 680 | ||
BC052341.1 | - | 6 | 784 | 5757011 | ||
D87470.1 | - | 7 | 674 | |||
EF363480.1 | - | 6 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
