siRNA Details
Gene Symbol: FAM168A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286050.1 | NP_001272979.1 | 8 | 975 | ||
NM_001286051.1 | NP_001272980.1 | 5 | 630 | |||
NM_015159.2 | NP_055974.1 | 7 | 948 | |||
XM_017017415.1 | XP_016872904.1 | 5 | 555 | |||
XM_017017416.1 | XP_016872905.1 | 5 | 390 | |||
GenBank mRNA | AK292054.1 | - | 7 | 838 | ||
BC052341.1 | - | 7 | 942 | 5757011 | ||
D87470.1 | - | 8 | 832 | |||
EF197985.1 | - | 5 | 539 | |||
EF363480.1 | - | 7 | 942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
