siRNA Details
Gene Symbol: ATP10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025153.2 | NP_079429.2 | 7 | 1401 | ||
XM_006714833.2 | XP_006714896.1 | 6 | 982 | |||
XM_011534468.2 | XP_011532770.1 | 6 | 958 | |||
XM_011534469.1 | XP_011532771.1 | 7 | 1222 | |||
XM_011534472.2 | XP_011532774.1 | 2 | 698 | |||
XM_017009252.1 | XP_016864741.1 | 6 | 958 | |||
XM_017009253.1 | XP_016864742.1 | 6 | 982 | |||
GenBank mRNA | AB018258.2 | - | 7 | 1228 | ||
AK025130.1 | - | 2 | 510 | |||
AK090832.1 | - | 6 | 1217 | |||
AK295081.1 | - | 5 | 1067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
