siRNA Details
Gene Symbol: C22orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302819.1 | NP_001289748.1 | 1 | 474 | ||
NM_015372.2 | NP_056187.1 | 2 | 265 | |||
GenBank mRNA | AJ704765.1 | - | 2 | 216 | ||
AK294232.1 | - | 1 | 444 | |||
AK315264.1 | - | 2 | 263 | |||
AL050256.1 | - | 2 | 253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
