siRNA Details
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 14 | 1924 | ||
XM_006718795.3 | XP_006718858.1 | 3 | 438 | |||
XM_011542691.2 | XP_011540993.1 | 15 | 2611 | |||
XM_011542692.2 | XP_011540994.1 | 15 | 2599 | |||
XM_011542694.2 | XP_011540996.1 | 15 | 2614 | |||
XM_011542695.2 | XP_011540997.1 | 14 | 2142 | |||
XM_017017393.1 | XP_016872882.1 | 14 | 2629 | |||
XM_017017394.1 | XP_016872883.1 | 15 | 2614 | |||
XM_017017395.1 | XP_016872884.1 | 15 | 2613 | |||
XM_017017396.1 | XP_016872885.1 | 8 | 1953 | |||
GenBank mRNA | AB028953.1 | - | 2 | 166 | ||
AK097578.1 | - | 11 | 1654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
