siRNA Details
Gene Symbol: UHRF1BP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006947.1 | NP_001006948.1 | 5 | 598 | ||
NM_015054.1 | NP_055869.1 | 5 | 598 | |||
XM_005268738.2 | XP_005268795.1 | 6 | 704 | |||
XM_005268739.3 | XP_005268796.1 | 5 | 558 | |||
XM_017019047.1 | XP_016874536.1 | 5 | 558 | |||
XM_017019048.1 | XP_016874537.1 | 6 | 704 | |||
XM_017019049.1 | XP_016874538.1 | 5 | 558 | |||
XM_017019050.1 | XP_016874539.1 | 5 | 558 | |||
XM_017019051.1 | XP_016874540.1 | 6 | 704 | |||
GenBank mRNA | AB014601.2 | - | 1 | 39 | ||
AK125813.1 | - | 5 | 558 | |||
BC014891.1 | - | 5 | 581 | |||
BC127017.1 | - | 5 | 416 | 40124660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
