siRNA Details
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 15 | 2204 | ||
XM_011537044.2 | XP_011535346.1 | 16 | 2306 | |||
XM_011537045.2 | XP_011535347.1 | 15 | 2116 | |||
GenBank mRNA | AK094697.1 | - | 14 | 2048 | ||
AL832314.1 | - | 14 | 5620 | |||
AL833497.1 | - | 3 | 1053 | |||
BC113389.1 | - | 14 | 1984 | |||
BC113391.1 | - | 14 | 1984 | |||
BC136775.1 | - | 15 | 2041 | 9020765 | ||
BC136776.1 | - | 15 | 2041 | 9020766 | ||
BC144407.1 | - | 16 | 2109 | 9052928 | ||
BX647116.1 | - | 15 | 2157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
