siRNA Details
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 15, 16 | 2230 | ||
XM_011537044.2 | XP_011535346.1 | 16, 17 | 2332 | |||
XM_011537045.2 | XP_011535347.1 | 15, 16 | 2142 | |||
GenBank mRNA | AK094697.1 | - | 14, 15 | 2074 | ||
BC113389.1 | - | 14, 15 | 2010 | |||
BC113391.1 | - | 14, 15 | 2010 | |||
BC136775.1 | - | 15, 16 | 2067 | 9020765 | ||
BC136776.1 | - | 15, 16 | 2067 | 9020766 | ||
BC144407.1 | - | 16, 17 | 2135 | 9052928 | ||
BX647116.1 | - | 15, 16 | 2183 | |||
L40392.1 | - | 1, 2 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
