siRNA Details
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 19 | 2952 | ||
XM_011537044.2 | XP_011535346.1 | 20 | 3054 | |||
XM_011537045.2 | XP_011535347.1 | 19 | 2864 | |||
GenBank mRNA | AK094697.1 | - | 18 | 2796 | ||
AL832314.1 | - | 17 | 6740 | |||
AL833497.1 | - | 6 | 2172 | |||
BC136775.1 | - | 19 | 2790 | 9020765 | ||
BC136776.1 | - | 19 | 2791 | 9020766 | ||
BC144407.1 | - | 20 | 2857 | 9052928 | ||
BX647116.1 | - | 19 | 2905 | |||
L40392.1 | - | 5 | 1036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
