siRNA Details
Gene Symbol: ATP13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020410.2 | NP_065143.2 | 24 | 3294 | ||
GenBank mRNA | AB058728.1 | - | 24 | 3264 | ||
AF288687.1 | - | 24 | 2912 | |||
AK026044.1 | - | 8 | 1444 | |||
AK056420.1 | - | 24 | 3270 | |||
AK074159.1 | - | 15 | 4079 | |||
AK074798.1 | - | 12 | 1799 | |||
AK074920.1 | - | 13 | 1874 | |||
AK095287.1 | - | 7 | 2467 | |||
AK123780.1 | - | 7 | 1638 | |||
AK125861.1 | - | 8 | 3591 | |||
AK130125.1 | - | 15 | 1934 | |||
AK172778.1 | - | 23 | 2725 | |||
AL834180.1 | - | 1 | 1743 | |||
BC009302.2 | - | 6 | 738 | |||
BC069211.1 | - | 5 | 628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
