siRNA Details
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 3, 4 | 551 | ||
NR_102264.1 | 4, 5 | 531 | ||||
XM_006717680.3 | XP_006717743.1 | 4, 5 | 931 | |||
GenBank mRNA | AK308333.1 | - | 3, 4 | 551 | ||
AK308358.1 | - | 1, 2 | 192 | |||
BC063664.1 | - | 3, 4 | 464 | |||
BC111740.1 | - | 4, 5 | 531 | 40038149 | ||
D42046.1 | - | 3, 4 | 493 | |||
DC404138.1 | - | 3, 4 | 552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
